2-[4-(Brommethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolan 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-(Bromomethyl)phenylboronic acid Pinacol ester, 4-Bromomethylphenylboronic acid, pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl bromide, 2-[4-(Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
L19954.03
L19954.06
L19954.03EA
98.8
EUR
L19954.03
L19954.06
2-[4-(Brommethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolan 95%
2-(4-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Purity: 95%
CAS No.: 138500-85-3
Molecular Formula: C13H18BBrO2
Formula Weight: 297.00
Melting Point: 83-85°
Beilstein Reference: 7921808
UN No.: UN3261
MDL No.: MFCD02179493
Formel:
C₁₃H₁₈BBrO₂ Schmelzpunkt: 83…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179493 CAS-Nummer: 138500-85-3 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...