Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate 97%

Lieferant: Thermo Fisher Scientific
Achtung

L19969.06
L19969.06EA 124 EUR
L19969.06
Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate 97%
Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate

Purity: 97% CAS No.: 91600-33-8 Molecular Formula: C7H11F3O3 Formula Weight: 200.16 Boiling Point: 180-184° Beilstein Reference: 6194166 MDL No.: MFCD00190634

Formel: C₇H₁₁F₃O₃
Siedepunkt: 180…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00190634
CAS-Nummer: 91600-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR