Ethyl-2,2-diethylbutyrate 98%
Lieferant: Thermo Scientific
L19972.06
L19972.06EA
198
EUR
L19972.06
Ethyl-2,2-diethylbutyrate 98%
Ethyl-2,2-diethylbutyrate
Purity: 98%
CAS No.: 34666-17-6
Molecular Formula: C10H20O2
Formula Weight: 172.27
Refractive Index: 1.4055
Boiling Point: 85-87°/30mm
Beilstein Reference: 1757834
MDL No.: MFCD00154424
Formel:
C₁₀H₂₀O₂ Molecular Weight: 172,27 g/mol Siedepunkt: 85…87 °C (30 mmHg) Dichte: 0,884 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00154424 CAS-Nummer: 34666-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...