Ethyl-(2-fluorphenyl)acetat ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl 2-fluorophenylacetate

L19999.06
L19999.06EA 36.8 EUR
L19999.06
Ethyl-(2-fluorphenyl)acetat ≥99%
Ethyl-(2-fluorphenyl)acetat

Purity: 99% CAS No.: 584-74-7 Molecular Formula: C10H11FO2 Formula Weight: 182.20 Flash Point: 85°(185°F) Refractive Index: 1.4820 Boiling Point: 235° MDL No.: MFCD04039343

Formel: C₁₀H₁₁FO₂
Molecular Weight: 182.20 g/mol
Siedepunkt: 235 °C
Dichte: 1.101
Flammpunkt: 85 °C (185 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD04039343
CAS-Nummer: 584-74-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR