2,6-Dichlor-3-(trifluormethyl)pyridin ≥98%

Lieferant: Thermo Scientific
Gefahr

L20003.03
L20003.03EA 141 EUR
L20003.03
2,6-Dichlor-3-(trifluormethyl)pyridin ≥98%
2,6-Dichloro-3-(trifluoromethyl)pyridine

Purity: 98% CAS No.: 55304-75-1 Molecular Formula: C6H2Cl2F3N Formula Weight: 215.99 Refractive Index: 1.4845 Boiling Point: 194-196° UN No.: 2810 MDL No.: MFCD00042245

Formel: C₆H₂Cl₂F₃N
Molecular Weight: 215,99 g/mol
Siedepunkt: 194…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042245
CAS-Nummer: 55304-75-1
EINECS: 259-585-8
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR