2-Acetamido-5-aminopyridin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-(Acetylamino)-5-aminopyridine
L20004.03
L20004.03EA
42.1
EUR
L20004.03
2-Acetamido-5-aminopyridin 98%
2-Acetamido-5-aminopyridin
Purity: 98%
CAS No.: 29958-14-3
Molecular Formula: C7H9N3O
Formula Weight: 151.17
Melting Point: 144-154°
Beilstein Reference: 128346
Formel:
C₇H₉N₃O Molecular Weight: 151,17 g/mol Schmelzpunkt: 144…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00160311 CAS-Nummer: 29958-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...