2-Brom-4-iodpyridin ≥97%

Lieferant: Thermo Scientific
Achtung

L20015.06
L20015.06EA 235 EUR
L20015.06
2-Brom-4-iodpyridin ≥97%
2-Brom-4-iodpyridin

Purity: 97% CAS No.: 100523-96-4 Molecular Formula: C5H3BrIN Formula Weight: 283.89 Melting Point: 61° Beilstein Reference: 108925

Formel: C₅H₃BrIN
Molecular Weight: 283,89 g/mol
Schmelzpunkt: 61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03085768
CAS-Nummer: 100523-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR