2-Brom-6-methylpyridin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L20018.06
L20018.14
L20018.06EA
82.2
EUR
L20018.06
L20018.14
2-Brom-6-methylpyridin ≥98%
2-Brom-6-methylpyridin
Purity: 98%
CAS No.: 5315-25-3
Molecular Formula: C6H6BrN
Formula Weight: 172.03
Flash Point: 97°(206°F)
Refractive Index: 1.5625
Boiling Point: 102-103°/20mm
Beilstein Reference: 107322
MDL No.: MFCD00040743
Formel:
C₆H₆BrN Siedepunkt: 102…103 °C (20 mmHg) Dichte: 1,512 Flammpunkt: 97 °C (206 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040743 CAS-Nummer: 5315-25-3 EINECS: 226-173-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...