3-Chlor-2-hydroxypyridin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Chloro-2-pyridinol

L20027.03 L20027.06 L20027.14
L20027.03EA 25.9 EUR
L20027.03 L20027.06 L20027.14
3-Chlor-2-hydroxypyridin ≥97%
3-Chlor-2-hydroxypyridin

Purity: 97% CAS No.: 13466-35-8 Molecular Formula: C5H4ClNO Formula Weight: 129.55 Melting Point: 180-182° Beilstein Reference: 109845

Formel: C₅H₄ClNO
Molecular Weight: 129.55 g/mol
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01646109
CAS-Nummer: 13466-35-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR