2,3,5-Trifluorisonicotinsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,3,5-Trifluoropyridine-4-carboxylic acid

L20050.06
L20050.06EA 231 EUR
L20050.06
2,3,5-Trifluorisonicotinsäure ≥97%
2,3,5-Trifluorisonicotinsäure

Purity: 97% CAS No.: 675602-91-2 Molecular Formula: C6H2F3NO2 Formula Weight: 177.08 Melting Point: 108-112° MDL No.: MFCD03840205

Formel: C₆H₂F₃NO₂
Molecular Weight: 177,08 g/mol
Schmelzpunkt: 108…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03840205
CAS-Nummer: 675602-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR