2,3,6-Trifluor-4-(trifluormethyl)pyridin ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
L20053.03
L20053.03EA
168
EUR
L20053.03
2,3,6-Trifluor-4-(trifluormethyl)pyridin ≥97%
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine
Purity: 97%
CAS No.: 84940-46-5
Molecular Formula: C6HF6N
Formula Weight: 201.07
Refractive Index: 1.3774
Boiling Point: 104-106°
UN No.: 1993
MDL No.: MFCD03840203
Formel:
C₆HF₆N Molecular Weight: 201,07 g/mol Siedepunkt: 104…106 °C Dichte: 1,579 Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840203 CAS-Nummer: 84940-46-5 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...