(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate 97%
Lieferant: Thermo Scientific
|
Gefahr
|
L20060.03
L20060.03EA
92.5
EUR
L20060.03
(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate 97%
(1S,2S)-(+)-2-Benzyloxycyclohexyl isothiocyanate
Purity: 97%
CAS No.: 737000-89-4
Molecular Formula: C14H17NOS
Formula Weight: 247.36
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5674
Melting Point: 33-36°
UN No.: UN2923
MDL No.: MFCD05664047
Formel:
C₁₄H₁₇NOS Molecular Weight: 247,36 g/mol Schmelzpunkt: 33…36 °C Dichte: 1,106 |
MDL:
MFCD05664047 CAS-Nummer: 737000-89-4 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...