2-Methoxy-3-pyridinboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Methoxypyridine-3-boronic acid
L20094.03
L20094.06
L20094.03EA
61.5
EUR
L20094.03
L20094.06
2-Methoxy-3-pyridinboronsäure ≥98%
2-Methoxy-3-pyridineboronic acid
Purity: 98%
CAS No.: 163105-90-6
Molecular Formula: C6H8BNO3
Formula Weight: 152.95
Melting Point: 136-138°
MDL No.: MFCD03411572
Formel:
C₆H₈BNO₃ Schmelzpunkt: 136…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411572 CAS-Nummer: 163105-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...