2-Chlor-4-(trifluormethyl)phenylboronsäure 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L20103.03
L20103.06
L20103.03EA
45.2
EUR
L20103.03
L20103.06
2-Chlor-4-(trifluormethyl)phenylboronsäure 96%
2-Chlor-4-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BClF₃O₂ Schmelzpunkt: 110…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02684337 CAS-Nummer: 254993-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...