2-Chlor-5-(trifluormethyl)phenylboronsäure 96%
Lieferant: Thermo Fisher Scientific
L20104.03
L20104.06
L20104.03EA
48
EUR
L20104.03
L20104.06
2-Chlor-5-(trifluormethyl)phenylboronsäure 96%
2-Chlor-5-(trifluormethyl)phenylboronsäure
Purity: 96%
CAS No.: 182344-18-9
Molecular Formula: C7H5BClF3O2
Formula Weight: 224.38
MDL No.: MFCD00797335
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...