4-Chlor-3-(trifluormethyl)phenylboronsäure 96%

Lieferant: Thermo Fisher Scientific
Achtung

L20105.03 L20105.06
L20105.03EA 60.8 EUR
L20105.03 L20105.06
4-Chlor-3-(trifluormethyl)phenylboronsäure 96%
4-Chlor-3-(trifluormethyl)phenylboronsäure

Purity: 96% CAS No.: 176976-42-4 Molecular Formula: C7H5BClF3O2 Formula Weight: 224.38 MDL No.: MFCD03094999

Formel: C₇H₅BClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03094999
CAS-Nummer: 176976-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR