2-Brom-4-fluorbenzonitril ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

L20115.06
L20115.06EA 42 EUR
L20115.06
2-Brom-4-fluorbenzonitril ≥98%
2-Brom-4-fluorbenzonitril

Purity: 98% CAS No.: 36282-26-5 Molecular Formula: C7H3BrFN Formula Weight: 200.01 Melting Point: 74-78° Beilstein Reference: 1940597 UN No.: 3439 MDL No.: MFCD00672924

Formel: C₇H₃BrFN
Schmelzpunkt: 74…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00672924
CAS-Nummer: 36282-26-5
EINECS: 252-949-7
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR