4-Aminopiperidin 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Piperidin-4-amine

L20127.06
L20127.06EA 72.1 EUR
L20127.06
4-Aminopiperidin 95%
4-Aminopiperidin

Purity: 98% CAS No.: 13035-19-3 Molecular Formula: C5H12N2 Formula Weight: 100.16 Flash Point: 46°(114°F) Refractive Index: 1.4910 Boiling Point: 65°/18mm Beilstein Reference: 1105 UN No.: 2733 MDL No.: MFCD02179399

Formel: C₅H₁₂N₂
Molecular Weight: 100,16 g/mol
Siedepunkt: 65 °C (18 mmHg)
Dichte: 0,954
Flammpunkt: 46 °C (114 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD02179399
CAS-Nummer: 13035-19-3
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR