4-Amino-2,3,5,6-tetrafluorbenzoesäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Amino-2,3,5,6-tetrafluorobenzoic Acid

L20128.03 L20128.06
L20128.03EA 44.7 EUR
L20128.03 L20128.06
4-Amino-2,3,5,6-tetrafluorbenzoesäure 98%
4-Amino-2,3,5,6-tetrafluorbenzoesäure

Purity: 98% CAS No.: 944-43-4 Molecular Formula: C7H3F4NO2 Formula Weight: 209.11 Melting Point: 182-184° Beilstein Reference: 2121117

Formel: C₇H₃F₄NO₂
Molecular Weight: 209.1 g/mol
Schmelzpunkt: 182…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007647
CAS-Nummer: 944-43-4
EINECS: 213-409-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR