2-Brom-4-fluor-1-iodbenzol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Brom-5-fluor-2-iodbenzol, 3-Brom-1-fluor-4-iodbenzol

L20144.06 L20144.03
L20144.06EA 54.5 EUR
L20144.06 L20144.03
2-Brom-4-fluor-1-iodbenzol ≥98%
2-Brom-4-fluor-1-iodbenzol

Purity: 98+% CAS No.: 202865-73-4 Molecular Formula: C6H3BrFI Formula Weight: 300.90 Flash Point: 110°(230°F) Refractive Index: 1.6290 Boiling Point: 224-225° Beilstein Reference: 6288420 MDL No.: MFCD00142868

Formel: C₆H₃BrFI
Siedepunkt: 224…225 °C
Dichte: 1,33
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00142868
CAS-Nummer: 202865-73-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR