4-Brom-2,3,5,6-tetrafluorbenzoylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-Bromo-2,3,5,6-tetrafluorobenzene-1-carbonyl chloride

L20153.06
L20153.06EA 152 EUR
L20153.06
4-Brom-2,3,5,6-tetrafluorbenzoylchlorid ≥98%
4-Brom-2,3,5,6-tetrafluorbenzoylchlorid

Purity: 98% CAS No.: 122033-54-9 Molecular Formula: C7BrClF4O Formula Weight: 291.43 Refractive Index: 1.5170 Boiling Point: 95-100° UN No.: 3265 MDL No.: MFCD03001173

Formel: C₇BrClF₄O
Molecular Weight: 291,43 g/mol
Siedepunkt: 95…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03001173
CAS-Nummer: 122033-54-9
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR