3-Buten-1-amin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L20157.03
L20157.06
L20157.03EA
113
EUR
L20157.03
L20157.06
3-Buten-1-amin 97%
3-Buten-1-amin
Purity: 97%
CAS No.: 2524-49-4
Molecular Formula: C4H9N
Formula Weight: 71.12
Flash Point: -5°(23°F)
Refractive Index: 1.4240
Odour: Amine-like
Boiling Point: 74-77°
Beilstein Reference: 1735803
UN No.: 2733
MDL No.: MFCD03425859
Formel:
C₄H₉N Siedepunkt: 74…77 °C Dichte: 0,777 Flammpunkt: –5 °C (23 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425859 CAS-Nummer: 2524-49-4 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...