1-Bromo-4-(tert-butoxy)benzene 98%
Lieferant: Thermo Scientific
|
Achtung
|
L20158.06
L20158.06EA
120
EUR
L20158.06
1-Bromo-4-(tert-butoxy)benzene 98%
1-Bromo-4-(tert-butoxy)benzene
Purity: 98%
CAS No.: 60876-70-2
Molecular Formula: C10H13BrO
Formula Weight: 229.12
Refractive Index: 1.5270
Boiling Point: 63-65°/0.4mm
Beilstein Reference: 2516411
MDL No.: MFCD00792676
Formel:
C₁₀H₁₃BrO Siedepunkt: 63…65 °C (0,4 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792676 CAS-Nummer: 60876-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...