2,2-Dimethyl-1,3-dioxan-5-on 90%, tech.

Lieferant: Thermo Scientific

L20177.03
L20177.03EA 196 EUR
L20177.03
2,2-Dimethyl-1,3-dioxan-5-on 90%, tech.
2,2-Dimethyl-1,3-dioxan-5-one

Purity: 90% CAS No.: 74181-34-3 Molecular Formula: C6H10O3 Formula Weight: 130.15 Storage Temperature: -30°C to -10°C Refractive Index: 1.4315 Boiling Point: 40-46°/14mm Beilstein Reference: 3061933 MDL No.: MFCD00671514

Formel: C₆H₁₀O₃
Molecular Weight: 130,14 g/mol
Siedepunkt: 40…46 °C (14 mmHg)
Lagertemperatur: Gefrierschrank
MDL: MFCD00671514
CAS-Nummer: 74181-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR