(1H-Imidazol-4-yl)methanol Hydrochlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Imidazolemethanol hydrochloride, 4(5)-Imidazolemethanol hydrochloride, 4-(Hydroxymethyl)imidazole hydrochloride
L20182.06
L20182.14
L20182.06EA
73.5
EUR
L20182.06
L20182.14
(1H-Imidazol-4-yl)methanol Hydrochlorid ≥98%
(1H-Imidazol-4-yl)methanol Hydrochlorid
Purity: 98+%
CAS No.: 32673-41-9
Molecular Formula: C4H6N2O.HCl
Formula Weight: 134.57
Storage Temperature: 2°C to 8°C
Melting Point: 104-108°
Beilstein Reference: 3562041
MDL No.: MFCD00012697
Formel:
C₄H₇ClN₂O Schmelzpunkt: 104…108 °C |
MDL:
MFCD00012697 CAS-Nummer: 32673-41-9 EINECS: 251-150-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...