(S)-(-)-1-(4-Fluorphenyl)ethylisocyanat 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L20231.03
L20231.03EA
116
EUR
L20231.03
(S)-(-)-1-(4-Fluorphenyl)ethylisocyanat 95%
(S)-(-)-1-(4-Fluorphenyl)ethylisocyanat
Purity: 95%
CAS No.: 745783-74-8
Molecular Formula: C9H8FNO
Formula Weight: 165.17
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4980
UN No.: 2206
MDL No.: MFCD05664076
Formel:
C₉H₈FNO Molecular Weight: 165.17 g/mol |
MDL:
MFCD05664076 CAS-Nummer: 745783-74-8 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...