Cer(IV)sulfat, wasserfrei 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cerdisulfat

L20254.36
L20254.36EA 782 EUR
L20254.36
Cer(IV)sulfat, wasserfrei 97%
Cer(IV)sulfat

Purity: 97% CAS No.: 13590-82-4 Molecular Formula: Ce(SO4)2 Formula Weight: 332.24 Merck Reference: 14,1990 UN No.: 3085 MDL No.: MFCD00148852

Formel: Ce(SO₄)₂
Molecular Weight: 332,24 g/mol
Schmelzpunkt: 195 °C
Dichte: 3,91
Lagertemperatur: Raumtemperatur
MDL: MFCD00148852
CAS-Nummer: 13590-82-4
EINECS: 237-029-5
UN: 3085
ADR: 5.1,III
Merck Index: 14,1990

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR