6-Methoxy-2-naphthylglyoxal hydrate 98% (Trockengewicht)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L20260.03
L20260.06
L20260.03EA
62.7
EUR
L20260.03
L20260.06
6-Methoxy-2-naphthylglyoxal hydrate 98% (Trockengewicht)
6-Methoxy-2-naphthylglyoxal hydrate
Purity: 98%
CAS No.: 1172293-10-5
Molecular Formula: C13H10O3.xH2O
Formula Weight: 214.22(anhy)
Melting Point: 125-130°
MDL No.: MFCD05664091
Formel:
C₁₃H₁₂O₄ Schmelzpunkt: 125…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664091 CAS-Nummer: 745783-88-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...