3-Methoxyphenylglyoxal hydrate 97% (Trockengewicht)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L20263.03
L20263.03EA
62.7
EUR
L20263.03
3-Methoxyphenylglyoxal hydrate 97% (Trockengewicht)
3-Methoxyphenylglyoxal hydrate
Purity: 97%
CAS No.: 32025-65-3
Molecular Formula: C9H8O3.xH2O
Formula Weight: 164.16(anhy)
Melting Point: 94-98°
Boiling Point: 95-98°/0.05mm
Beilstein Reference: 1935225
MDL No.: MFCD05664093
Formel:
C₉H₈O₃·xH₂O Siedepunkt: 95…98 °C (0,05 mmHg) Schmelzpunkt: 94…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664093 CAS-Nummer: 32025-65-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...