2-Chlor-3-pyridinboronsäure 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Chlor-3-pyridinylboronsäure, 2-Chlorpyridin-3-boronsäure
L20303.03
L20303.03EA
36.4
EUR
L20303.03
2-Chlor-3-pyridinboronsäure 96%
2-Chloro-3-pyridineboronic acid
Purity: 96%
CAS No.: 381248-04-0
Molecular Formula: C5H5BClNO2
Formula Weight: 157.37
Melting Point: 160°
Beilstein Reference: 9119913
Formel:
C₅H₅BClNO₂ Molecular Weight: 157,36 g/mol Schmelzpunkt: 160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094997 CAS-Nummer: 381248-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...