5-tert-Butyl-2-methoxybenzeneboronic acid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (5-(tert-Butyl)-2-methoxyphenyl)boronic acid, 5-(tert-Butyl)-2-methoxyphenylboronic acid

L20415.03
L20415.03EA 102 EUR
L20415.03
5-tert-Butyl-2-methoxybenzeneboronic acid ≥98%
5-tert-Butyl-2-methoxybenzeneboronic acid

Purity: 98+% CAS No.: 128733-85-7 Molecular Formula: C11H17BO3 Formula Weight: 208.07 Melting Point: 103-106° Beilstein Reference: 4181890 MDL No.: MFCD06201034

Formel: C₁₁H₁₇BO₃
Molecular Weight: 208,07 g/mol
Schmelzpunkt: 103…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06201034
CAS-Nummer: 128733-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR