LR white acrylic resins, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ200

L9774-100G L9774-500G
L9774-100GEA 143 EUR
L9774-100G L9774-500G
LR white acrylic resins, Sigma-Aldrich®
LR white acrylic resins

Benzoyl peroxide (B5907) is specifically formulated as an initiator for these resins.

Lagertemperatur: Kühlschrank
MDL: MFCD00131541
CAS-Nummer: 94188-59-7

Bestellen

Spezifikation Testergebnisse

Refractive Index
at 25 ° C: 1.4592 (white light 20 deg C)
Conforms
Cure Time
at 25 deg C (Benzoyl Peroxide 1% w.v)
≤20 min


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR