ClipTip™ Racks ohne Spitzen
Lieferant: Thermo Fisher Scientific
94410519 94410819 94410219 94410619
613-2221EA 22.4 EUR
613-2221 613-2226 613-2227 LABS94410619
ClipTip™ Racks ohne Spitzen
Racks und Boxen für Pipettenspitzen
Utilize Thermo Scientific™ ClipTip™ Pipette tip Racks, available in three sizes.
  • Small rack: For use with ClipTip™ 20 and 50 Pipette tips
  • Medium rack: For use with ClipTip™ 200 and 300 Pipette tips
  • Large rack: For use with ClipTip™ 1000 Pipette tips
Bestellinformation: Racks are supplied without tips and inserts.