Rechargeable safety helmet torch, rechargeable, ADALIT® L-10
Lieferant: LACONT UMWELTTECHNIK
B69-7200-A
LACOB69-7200-AEA 0 EUR
LACOB69-7200-A
Rechargeable safety helmet torch, rechargeable, ADALIT® L-10
Beleuchtung Taschenlampen
The ADALIT L-10 is a high performance rechargeable LED safety torch designed specifically for firefighting helmets. ADALIT L-10 is a powerful hands-free torch that also incorporates a beacon to signal the position of firefighter.
  • High intensity LED white lighting
  • Ultra-lightweight design
  • Firefighter locator using a high intensity red LED flashing beacon
  • Lithium-ion rechargeable battery with indication of remaining charge and battery level
  • Manufactured from thermoplastic resin for high impact resistance and protection
Dimension: 156×56×48 mm
Weight: 180 g