Safety storage cabinets for hazardous substances, type 30
Lieferant: LACONT UMWELTTECHNIK
B80-2600-A
LACOB80-2600-AEA 0 EUR
LACOB80-2600-A
Safety storage cabinets for hazardous substances, type 30
Möbel Lagerschränke
Cabinet body made of B1 material RAL 7035 - light grey and doors either in RAL 7035 - light grey or RAL 1007 - daffodil yellow. It has fire resistance up to 30 minutes.
  • Height-adjustable shelves at intervals of 32 mm
  • Automatic thermal closing mechanism if fire occurs
  • DN 80 ventilation connection for exhaust air extraction on cabinet roof
  • Leveling feet for installing cabinet horizontally
Standards: EN 14470-1 and TRbF 20 - L