Safety cabinets for gas cylinders, FCG
Lieferant: LACONT UMWELTTECHNIK
C27-3101-B C27-3131-B C27-3134-B C27-3133-B C27-3132-B C27-3136-B C27-3102-B C27-3105-B C27-3135-B C27-3106-B C27-3103-B C27-3104-B C273110-B C27-3113-B C27-3112-B C27-3119-B C27-3121-B C27-3118-B C27-3120-B
LACOC27-3101-BEA 0 EUR
LACOC27-3101-B LACOC27-3131-B LACOC27-3134-B LACOC27-3133-B LACOC27-3132-B LACOC27-3136-B LACOC27-3102-B LACOC27-3105-B LACOC27-3135-B LACOC27-3106-B LACOC27-3103-B LACOC27-3104-B LACOC273110-B LACOC27-3113-B LACOC27-3112-B LACOC27-3119-B LACOC27-3121-B LACOC27-3118-B LACOC27-3120-B
Safety cabinets for gas cylinders, FCG
Möbel Lagerschränke
Gas cylinder cabinets and containers are intended for outdoor use and for an outdoor storage facility.
  • Welded profile steel construction
  • Grid-type door
  • Cantilever arm compartment divider with safety chains
  • Lifting attachment for cranes as standard
  • Single or double door depending on version
Side walls and roof made from galvanised and painted sheet steel whereas back wall made from galvanised and coated trapezoidal sheet.
Standards: Conforms with TRGS 510.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable