Safety storehouses with natural ventilation for hazardous substances, SLH-NB
Lieferant: LACONT UMWELTTECHNIK
H63-1104-A H63-2104-A H63-1100-A H63-2100-A H63-1101-A H63-2101-A H63-1102-A H63-1109-C H63-2102-A H63-2109-C
LACOH63-1104-AEA 0 EUR
LACOH63-1104-A LACOH63-2104-A LACOH63-1100-A LACOH63-2100-A LACOH63-1101-A LACOH63-2101-A LACOH63-1102-A LACOH63-1109-C LACOH63-2102-A LACOH63-2109-C
Safety storehouses with natural ventilation for hazardous substances, SLH-NB
Möbel Lagerschränke
Safety storehouses are ideal for passive storage of flammable liquids.
  • Ground connection to avoid dangerous electrostatic charges
The legally required 0,4-fold minimum air exchange rate is achieved by continuous ventilation slots directly above the base sump tray and ventilation slots in the upper area of back wall and door.
Standards: Rate of air exchange has been tested and approved in accordance with the latest requirements of Z-38,5-171 DIBt Berlin and STATIC approved.