Sumps for small containers, PE-EP
Lieferant: LACONT UMWELTTECHNIK
P70-7083-A
LACOP70-7083-AEA 0 EUR
LACOP70-7083-A
Sumps for small containers, PE-EP
Fasslager
Ideal for the storage of water-endangering and aggressive media.
  • Rotationally-moulded body of red PE
  • Circumferential stabilising ribs
  • Equipped with protective cover