Polyethylene Heavy-Duty Buckets with Spout
Lieferant: LAMAPLAST
1140 1141N 1140N 1141 1142N 1142
LAMA1140EA 17.3 EUR
LAMA1140 LAMA1141N LAMA1140N LAMA1141 LAMA1142N LAMA1142
Polyethylene Heavy-Duty Buckets with Spout
Eimer
Made with low density polyethylene (LDPE), in white or black colour options.
  • Conical shape, litre graduations
  • With ergonomic handle
  • Large spout for easy pouring
  • Food contact material approved for white models
Buckets feature a flat bottom design to ensure total emptying and allow for easy cleaning.