Square Buckets
Lieferant: LAMAPLAST
1137 1147
LAMA1137EA 69.6 EUR
LAMA1137 LAMA1147
Square Buckets
Eimer
Robust PP bucket with ergonomic handle.
  • Square shape
  • Flat bottom ensures total emptying and easy cleaning
  • Food contact material approved
Hot filling possible up to 95 °C.