Garbage Bins with Lids
Lieferant: LAMAPLAST
1152BL
LAMA1152BLEA 166 EUR
LAMA1152BL
Garbage Bins with Lids
Behälter Abfallbehälter
Made of HDPE
  • White body and coloured lid
  • Good chemical resistance
Bins with floating lid.