Trays, rectangular
Lieferant: LAMAPLAST
1631
LAMA1631EA 40.8 EUR
LAMA1631
Trays, rectangular
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
PS, white.
  • Stable, good break-resistance
  • Temperature resistant from −178 to 80 °C (melamine) and −40 to 55 °C (PS)
  • Smooth surfaces for easy cleaning
Multipurpose dishes for a wide variety of laboratory applications. Also suitable for use in the food industry.