Transparent Storage Box with Lid
Lieferant: LAMAPLAST
1909
LAMA1909EA 90.1 EUR
LAMA1909
Transparent Storage Box with Lid
Boxen Lagerboxen
Space saving container. Stackable with lid and nestable when not in use.
  • Suitable for food use
  • Perfect for goods handling
Ideal for long term storage.
Standards: Meets FCM regulations 1935/2004/CE and UE 10/2011