Drums, narrow neck
Lieferant: LAMAPLAST
207
LAMA207EA 96.4 EUR
LAMA207
Drums, narrow neck
Fässer, Fässer, Trommeln
HDPE, narrow neck
  • Good chemical resistance
  • Good temperature resistance
  • Stackable
  • Self sealing caps with total polyurethrane foam gasket
Ideal for use with foods.