Drum, stackable
Lieferant: LAMAPLAST
5670
LAMA5670EA 105 EUR
LAMA5670
Drum, stackable
Fässer, Fässer, Trommeln
UN approved drum, HDPE, blue with screw cap.
  • Rectangular cross-section drum is ideal for palletising on pallets 1000×1200
  • Stackable
  • Self-sealing cap with total PE foam gasket
  • Food contact material approved according EU10/2011
Ideal for use with foods.