Plastic Crate with Lid
Lieferant: LAMAPLAST
693
LAMA693EA 175 EUR
LAMA693
Plastic Crate with Lid
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Crates have one handle on the longest side, this is placed centrally. Crates are light yet strong, providing an economical alternative to metal crates.
  • Polypropylene
  • Colour: Grey
  • For food use