Cooling Boxes
Lieferant: LAMAPLAST
783
LAMA783EA 344 EUR
LAMA783
Cooling Boxes
Boxen Kühl- und Gefrierboxen, Kühl- und Gefrierbeutel
Box made of PP, blue, with white lid.
  • Polyurethane thermal insulation between the walls of the container
  • Ideal for transporting substances to maintain a constant temperature
  • Suitable for use with foods