Culture flasks with baffles
Lieferant: LAT - Labor- & Analysen-Technik GmbH
192010024 192010044
LATA192010024EA 40.2 EUR
LATA192010024 LATA192010044
Culture flasks with baffles
Kolben Kulturkolben
Culture flasks, made of borosilicate glass, for strong turbulence when stirring in Erlenmeyer flasks.
  • For Kapsenberg caps
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes