Eclipse™ Gel Loading Capillary Pipette Tip, 200 µl
Lieferant: LABCON
1034-965-008-9
LBCN1034-965-008-9EA 2310 EUR
LBCN1034-965-008-9
Eclipse™ Gel Loading Capillary Pipette Tip, 200 µl
Pipettenspitzen
Eclipse™ tips are made of high quality 100% pure virgin medical grade polypropylene so there won't be recycled plastics in our tips. Resin in pre-tested for metal contamination and to ensure no extractables are present.
  • With gel loading capillary
  • Fits most popular 200 µl pipettors
  • ATP, RNase/DNase and endotoxin free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Autoclavable
  • Height: 82,5 mm
  • Made in USA
Eclipse™ tips in racks are packed in Teal racks. These racks are made of polypropylene and are autoclavable repeatedly. They have sturdy latch hinges that won't come apart like lesser quality tip racks and a special snap lock designed so that it will not catch on gloves.
Eclipse™ tips can be also got in resealable zip style bags, in refills for our Pagoda® system, in Eclipse™ refills, and in racks that have been sterilised using irradiation.
Standards: Every lot is lab tested and certified free of detectable Nuclease (Rnase/Dnase), Endotoxin (pyrogen) contamination and certified to U.S.P. results.