Eclipse Graduated Pipet Tips with UltraFine Point, 10 µl
Lieferant: LABCON
1038-960-038-9
LBCN1038-960-038-9EA 848 EUR
LBCN1038-960-038-9
Eclipse Graduated Pipet Tips with UltraFine Point, 10 µl
Pipettenspitzen
Eclipse 10 µl Graduated Pipet Tips with UltraFine Point are precision-engineered by Labcon in the USA. Each case contains 11,520 tips conveniently arranged in 96 racks.
The ultrafine point ensures accurate and consistent liquid handling, making them ideal for various laboratory applications requiring precision and reliability. These pipet tips are graduated for quick volume identification and are designed to fit seamlessly with a wide range of pipettors. The high-quality polypropylene construction guarantees excellent clarity and chemical resistance, safeguarding sample integrity during experiments.
With Eclipse 10 µl Graduated Pipet Tips, researchers can streamline their workflows and achieve reproducible results, ultimately enhancing overall lab efficiency. Trust in Labcon's expertise for dependable laboratory consumables that meet the rigorous demands of scientific research.